Polypropylene Cylinders
Supplier: Bel-Art
28458-6000_SE 470148-670 470148-836 470177-816 470177-818 470188-958 470188-960
470158-130CS 526.15 CAD
470158-130 470148-670 470148-836 470177-816 470177-818 470188-958 470188-960
Polypropylene Cylinders
Cylinders Measuring Cylinders
Unbreakable Polypropylene.
  • Permanent, molded, single scale metric markings
  • Flared top rims and large spouts facilitate pouring
  • Hexagonal shaped bases prevent tipping and rolling
  • Autoclavable at 121°C
Chemically resistant to most acids, bases, alcohols, esters, and ketones.
Order Now
SPECIFICATIONS
Environmentally Preferable