Medullated Nerve, Cross/Longitudinal Section, Masson Stained Slides
Supplier: Avantor
470182-696EA 11.95 CAD
470182-696
Medullated Nerve, Cross/Longitudinal Section, Masson Stained Slides
Slides Prepared Slides
Demonstrates a Variety of Nervous Tissue
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Mammal. Peripheral nerve bundles showing axis cylinders, medullary sheaths, Schwann nuclei, epi- and perineurium.