Ward's® Live Pseudomonas aeruginosa culture (Pathogenic)
Supplier: Avantor
470179-204EA 22.05 CAD
470179-204 470179-540
Ward's® Live Pseudomonas aeruginosa culture (Pathogenic)
Specimens Live Specimens Bacteria - Living Material
This organism can be isolated from many human urinary tract infections, as well as polluted water and sewage. It is commonly identified by its distinctive grape-like smell (trimethylamine) or because it can turn the agar light green due to production of a blue pigment (pyocyanin).
  • Biosafety Level: 2
  • Canada: Containment Level 2 - PHAC Licence required
  • Gram Stain: Negative
  • Morphology: rods
  • Motility: Positive
  • Respiration: Aerobic
  • Chromogenesis: Yes (blue)
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, soMore Product Information