Ward's® Live Staphylococcus saprophyticus Culture
Supplier: Avantor
470179-162EA 22.05 CAD
470179-162 470179-578
Ward's® Live Staphylococcus saprophyticus Culture
Specimens Live Specimens Bacteria - Living Material
Grows on dead tissues. Novobiocin resistant (S. epidermidis and S. aureus novobiocin sensitive). 
  • Biosafety Level: 1
  • Canada: Containment Level 2 - PHAC Licence required
  • Coagulase: Negative
  • Catalase: Positive
  • Motility: Negative
  • Morphology: Cocci in clusters
  • Gram reaction: Positive
  • Respiration: Aerobic
All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
This organism is offered as a ready-to-use tube culture, as well as in aMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable