Student-Grade Polypropylene Beakers
Supplier: Shiv Dial Sud & Sons
CH10155/1 F26212-0000_SE BP0600 F26213-0000 F26214-0000 CH10155/3
470178-270CS 422.4 CAD
470178-270 470178-272 470178-274 490012-412 470178-269 470222-440
Student-Grade Polypropylene Beakers
Beakers
These simple, stackable, ribbed beakers with raised and permanent graduations are an excellent value.
  • Polypropylene construction
  • Stackable
  • Simple to use
  • Permanent graduations for easy measurements
  • Autoclavable at 121 °C
Perfect for use with acids, bases, and alcohols, they can also be used short-term with hydrocarbons.