Ward's® Hay Medium Solution
Supplier: Avantor
470177-390PK 39.5 CAD
470177-390 470180-766
Ward's® Hay Medium Solution
Cell Culture Supplements and Additives
Similar medium to Similar to Cereal Grass medium and Cerophyll. For culture of protists.
  • Sterile- no autoclaving
  • Convenient - prepared for you
  • Prepared consistenly every time
Concentrate comes with a vial of heat-treated wheat seeds.
Ordering information: These products contain living or perishable materials
Delivery information: Specimens will ship via overnight or 2nd day delivery. Please specify a delivery date during checkout.
Order Now
SPECIFICATIONS
Environmentally Preferable
Species
Source
Learn more