Ward's® Live Bacillus subtilis Culture
Supplier: Avantor
470176-524EA 22.05 CAD
470176-524 470176-526
Ward's® Live Bacillus subtilis Culture
Specimens Live Specimens Bacteria - Living Material
This organism is commonly isolated in soil and produces antibiotics such as bacitracin, subtilisin, and mycobacillin.
  • Biosafety Level: 1
  • Gram Stain: Positive
  • Morphology: rods in chains
  • Motility: Positive
  • Respiration: Aerobic
  • Spores: Yes, centrally located
All bacterial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.
It's gram stain is positive, and its spores are centrally located.
Ordering information: This organism is offered as a ready-to-use tubeMore Product Information