Ward's® Live Halobacterium salinarium Culture

Supplier: AMERICAN TYPE CULTURE COLL.
19700
470176-326EA 22.05 CAD
470176-326
Ward's® Live Halobacterium salinarium Culture
Specimens Live Specimens Bacteria - Living Material
Red-pigmented halophile from evaporating salt ponds; requires 25% salt medium, lyses easily in plain water, releasing DNA.

  • Biosafety Level: 1
  • Environment: High Salt (halophile)
  • Motility: Positive
  • Morphology: Variable (rods in 25% salt, coccal in less than 15% salt)
  • Gram Stain: Negative but difficult to see due to salt requirement
  • Respiration: Aerobic

All microbial cultures undergo an extensive quality control program prior to releasing for sale, so they are guaranteed for purity as well as for species.

Halophilic organisms like this one require aMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR