Premium Bosshead
Supplier: Wards
PRBH02
470165-424EA 20.05 CAD
470165-424
Premium Bosshead
Clamps Bossheads
Great for Connecting Rods and Apparatus in Tight Places without Sacrificing Strength
  • Threaded metal one-piece clamping screws
  • Able to hold rods up to 15 mm in diameter
  • Holds rods orthogonal (90°) to one another
  • All metal construction to carry heavy loads and prevent torque damage
Order Now
SPECIFICATIONS
Environmentally Preferable