Premium Two-Prong Clamp, Vinyl Coated
Supplier: Wards
PRCMP05
470165-410EA 43.6 CAD
470165-410
Premium Two-Prong Clamp, Vinyl Coated
Clamps General Purpose Clamps
Vinyl Lined Extension Lab Clamp
  • Maximum clamp opening is 11 cm
  • 14 cm connecting rod.
Clamp constructed of powder coated steel, two vinyl coated prongs attached to an aluminum rod. Completely metal construction with thread fitted actuating joints and 10 mm diameter connecting rod.