Forensic Chemistry: Chemical Detection of Fingerprints Kit
Supplier: Ward's Science
IS9013
470163-330KT 95.65 CAD
470163-330
Forensic Chemistry: Chemical Detection of Fingerprints Kit
Educational Materials Biology Educational Materials Blood Typing Learning Activities
Identifying Fingerprints Without Powder!
  • Three activities in one convenient kit
  • All required chemicals are included
Utilize alternative methods for detecting fingerprints. Examine some possible methods of gathering evidence when dusting for fingerprints is not effective. Learn to identify fingerprint types, a method of fuming for fingerprints, and a technique of chemically-developing fingerprints. The three activities include fingerprint analysis, ninhydrin development, and cyanoacrylate fuming.
Ordering information: Includes Super Bonding Glue,More Product Information