WaterWorks™ School Test Kit
Supplier: Industrial Test Systems
487995 487942
470004-284EA 261.8 CAD
470004-284 470151-358
WaterWorks™ School Test Kit
Educational Materials Earth Science Educational Materials Natural Resources Learning Activities
The WaterWorks School Test Kit is a great introduction to water-quality testing.
  • Safe, easy-to-use strips
  • Nine different tests
  • Enough material to test each factor 60 times
  • Manual included
  • Great introduction to water-quality testing
These safe, easy-to-use test strips eliminate the need to mix and dispose of hazardous chemicals and handle glassware. You just dip a test strip in a water sample and then compare it to the included color chart to interpret the results. This kit contains tests for free chlorine, total chlorine, iron, copper, pH, nitrates,More Product Information