Crucible Holder
Supplier: Plasticoid
VCHWGS
470151-210CS 161.5 CAD
470151-210
Crucible Holder
Supports Support Sleeves
Made of molded, black rubber with a conical opening 27 mm at the top tapered to 22 mm at the bottom, the crucible holder includes a glass funnel.
  • Holder may be used with suction flasks of 250 to 1000 ml capacity
  • Holds Gooch and similar crucibles of 35 to 40 mm diameter at the top
Overall length: 60 mm.