Braun Electroscope with Accessories
Supplier: Shiv Dial Sud & Sons
P45262
470149-478CS 2943 CAD
470149-478
Braun Electroscope with Accessories
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Versatile Apparatus for Electrostatics Lessons.
  • Includes instructions
This rugged, sensitive electroscope can accept various accessories to change its capacitance. A sensitive aluminum vane is balanced in a vertical position on a metal support which is insulated from its base. The vane is easily observed and projected on a chalkboard (Light Source, not included), on which an arbitrary scale has been marked. Height without attachments is about 9". Four attachments, each equipped with a banana plug which fits the vane system top, are included; a 3"More Product Information