Ceramic Ring Magnets
Supplier: Shiv Dial Sud & Sons
P40077/6
470149-462CS 3790.3 CAD
470149-462
Ceramic Ring Magnets
Magnets
Ceramic Ring Magnets have a high resistance to demagnetization and don't need a keeper.
  • High Resistance to Demagnetization
  • No Keeper Needed
  • Magnets Demonstrate Repulsion
  • Barium and Iron Oxide Construction
  • Width: 6 mm
They're made of a mixture of barium and iron oxides and are magnetized along their length. The central hole allows them to be threaded onto a pencil or glass rod to demonstrate repulsion.
They may be stacked four-to-five high this way. Each magnet is 6 mm thick and is available in two sizes.