Ice Melting Blocks
Supplier: Shiv Dial Sud & Sons
PH20050A
470148-814CS 1777.5 CAD
470148-814
Ice Melting Blocks
Educational Materials Physics Educational Materials Heat/Thermodynamics Learning Activities
Ice Melting Blocks are a unique way to teach principles of conductivity of heat and the scientific method.
  • Instructions Included
  • Two Blocks Look Similar but Have Different Construction
  • One Block Seems Cold, One Block Seems Hot
  • Students Learn Principles of Conductivity
  • Demo Uses Scientific Method
They consist of two blocks of similar appearance but composed of different materials. One block seems cold to the touch, and the other seems warm.
Teachers ask students if they think it would take longer for an ice cube to melt on the cold block. TeachersMore Product Information