Funnel Set, Dynalon
Supplier: Dynalon
241645-0SET
470146-840KT 133.15 CAD
470146-840
Funnel Set, Dynalon
Funnels Filtration Funnels Filter Funnels
Two each of top diameter 65mm and 100mm and 120mm. All are 60° angle. Set of six.