Stopcock/Vacuum Grease
Supplier: Liveo
1597418
470146-194CS 1029.25 CAD
470146-194
Stopcock/Vacuum Grease
Lubricants
Highly Resistant to Water, Alkalis, and Chemicals
  • Silicone based lubricant for general use
  • Sealant for vacuum and pressure systems
  • Has a low volatility
  • Withstands temperatures from -40 to 260°C