TeacherGeek Motor Adaptor Pin
Supplier: TEACHERGEEK INC
1823-37
470132-442EA 12.15 CAD
470132-442
TeacherGeek Motor Adaptor Pin
Educational Materials Physics Educational Materials TeacherGeek System Components
TeacherGeek Motor Adaptor Pins slide onto the motor shaft and fit securely in the splined hole of a small gear or pulley.
This allows you to easily motorize an activity or original design.