Electric Heaters
Supplier: Glo-Quartz
LHP-200
470122-462EA 143.9 CAD
470122-462 470122-464
Electric Heaters
Heaters Hot Plates
Excellent for heating all acid solutions except hydrofluoric.
  • Resistance element is completely sealed in a quartz tube
  • Upper portion remains cool and can be held in the bare hand
  • Diameter: ⁵/₁₆"
Permanently attached 6' 2-wire line cord/plug for 115 VAC or DC.