Large Tinted Desiccator Cabinet
Supplier: Bel-Art
H42053-0001_SE
470121-956EA 1143.9 CAD
470121-956
Large Tinted Desiccator Cabinet
Desiccators Desiccator Cabinets
Made of smoke tinted glass. Door has top single roll latch. Four sets of shelf rails allow for low or high objects. Includes small tray for silica gel and two vented shelves. InsideDimensions: 9" L x 8" D x 9" H (23 x 23 x 20 cm).Overall dimensions: 10" L x 10" D x 10" H (25 cm).