Principles and Practice of Agarose Gel Electrophoresis Kit
Supplier: Edvotek
101
470121-714EA 151.5 CAD
470121-714
Principles and Practice of Agarose Gel Electrophoresis Kit
Educational Materials Chemistry Educational Materials Chromatography Learning Activities
The Principles and Practice of Agarose Gel Electrophoresis Kit demonstrates how electrophoresis separates molecules on the basis of size and charge.
  • Safe, Colorful, and Fast
  • Time Required: 45 Minutes
  • Kit Demonstrates Electrophoresis
  • Instructions Included
  • Ready-To-Load Dye Samples Included
It's a safe, colorful, fast, and simple way to teach a technique that engages students.
The activity requires approximately 45 minutes.
Ordering information: This kit includes instructions, Ready-to-Load dye samples, agarose powder, practice gel-loadingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable