Eisco® NextGen Bunsen Burner with Flame Stabilizer
Supplier: Wards
CH0992NG CH0992LP
470121-694EA 53.7 CAD
470121-694 470121-698
Eisco® NextGen Bunsen Burner with Flame Stabilizer
Heaters Gas Burners
The next generation in laboratory burners. NextGen Bunsen Burners are designed to be the safest on the market.
  • Designed for safety
  • Expanded cast iron base prevents easy tipping, even with hoses connected
  • Spaced feet so burner fits comfortably under stands
  • Candle stick style handle allows user to move burner without getting harmed
  • Features air flow adjustment sleeve and brass flame stabilizer
  • Base is coated with a chemical resistant powder coating
This basic Bunsen burner produces a safe continuous stream of flammable gas used for heating,More Product Information