Decade Inductance Box
Supplier: Wards
PH1135F
470121-490EA 393.35 CAD
470121-490
Decade Inductance Box
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Investigating Inductance has never been this easy
This sturdy decade box allows a range of inductances from 10 H down to 1 microH. Maximum capacity is 11.11111 H. Housed in a box with seven dials and fitted with 4 mm output plugs, the unit is fully equipped for laboratory use.