Simulaids® Water Rescue Manikins
Supplier: Nasco
149-1434 149-1435 149-1350 149-1329 149-1351 149-1352 1005765 149-1328
470119-962EA 3660.95 CAD
470119-962 470119-964 470119-966 470119-970 470119-972 470119-976 470119-978 470119-980 470119-982 470119-984
Simulaids® Water Rescue Manikins
Models Human Biology Models
Practice Water Rescues And Retrievals.
  • Articulated joints allow varying positions
  • Manikins will fully or partially submerge when filled with water
These Rescue manikins allow Coast Guards, Lifeguard Trainers and Emergency Personnel to train water rescue and retrieval as well as CPR. These manikins have articulated joints and are made of durable plastic and rust-resistant steel.
When filled with water the adult manikin will submerge to the neck, if an additional 3 or 5 kg (a brick will do) is added, it will sink completely. Child, infant and newbornMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable