Eco RI—
Supplier: Edvotek
715-B
470111-874EA 96.85 CAD
470111-874
Eco RI—
Enzymes
Includes instructions for rehydration.Recognition Site: 5'-G^AATTC-3' (1500 units) 150 ul—for 30 DNA digests
Order Now
SPECIFICATIONS
Enzyme name Eco RI—
Environmentally Preferable
Source
Species
Learn more