E.I. Double Convex Glass Lenses
Supplier: Wards
PH0534C-CV PH0534D-CV PH0534F-CV PH0534B-CV PH0534I-CV PH0534A-CV PH0534G-CV PH0534H-CV PH0534K-CV PH0534L-CV PH0534L-CV2 PH0534L-CV1 PH0534J-CV
470014-576EA 10.1 CAD
470014-576 470014-578 470014-580 470014-716 470017-098 470017-124 470111-256 470111-258 470111-262 470111-264 470135-008 470135-010 470105-726
E.I. Double Convex Glass Lenses
Educational Materials Physics Educational Materials Optics Learning Activities
Individual spherical glass lenses.
  • Double convex lenses in a variety of diameters and focal lengths
  • Made of high quality, optically worked glass
  • Highly polished surface, ground edges
This optically true lens is excellent for physics classroom demonstrations in optics and light refraction.
Eisco labs proudly manufactures its own products using state-of-the-art equipment, and following the highest standards of quality control.
Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality andMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable