Kemtec® Introduction To Plant Cloning
Supplier: Kemtec
2-504
470111-206KT 528.1 CAD
470111-206
Kemtec® Introduction To Plant Cloning
Educational Materials Biology Educational Materials Plant Learning Activities
Learn to clone plants.
  • Practice sterile technique
  • No hood or incubator required
Cloning plant tissue makes an excellent introduction to biotechnology and sterile technique as plant propagation by cloning becomes increasingly important in food and flower production. Students germinate carrot seeds in a sterile environment to provide plant material from which to establish a culture, use sterile technique to grow seedlings and establish a callus culture, and induce the development of multiple shoots (plantlets) from callus tissue. The process takesMore Product Information