Wave Demonstrations Set
Supplier: United Scientific Supplies
WDSET1 FCS003
470110-488EA 50.65 CAD
470110-488 470349-656
Wave Demonstrations Set
Educational Materials Physics Educational Materials Waves and Oscillations Learning Activities
This set provides versatile tools for a variety of demonstrations.
  • Set Provides Wide Range of Ways to Show Waves
  • Teacher's Manual Included
  • Wave Characteristics Include Pulse, Shape, Superposition, and Reflection
  • Two Springs Included
  • Waves Length Changes When Springs Are Connected
Students can explore a variety of wave characteristics including pulse, shape, superposition, and reflection.
A 7 cm dia. slinky and a 2 cm dia. spring coil exhibit different wave characteristics when used separately. However, when linked with the provided cotton cord,More Product Information