Benchmark TC 9639 Thermal Cyclers
Supplier: Ward's
T5000-384 T5000-96 T5000-A-IS
470107-136EA 13006.09 CAD
470107-136 470107-314 470107-352
Benchmark TC 9639 Thermal Cyclers
Thermal Cyclers
High quality thermal cyclers for laboratories with medium to high throughput needs.
  • Intuitive touchscreen control
  • Program wizard for quick, easy protocol entry
  • Multiblock version accommodates most sample sizes
  • 384 well block version available
  • Two year warranty
Designed for laboratories with medium to high throughput needs, Benchmark's Thermal Cyclers provide consistent, reliable results
A large, full color touch screen serves as the operations center where all functions are accessed. Starting at the “Home” screen, simply touch an icon to view theMore Product Information