Ward's® Rock Thin Section
Supplier: Avantor
470106-374KT 653.65 CAD
470106-374
Ward's® Rock Thin Section
Specimens Rock Specimens Rock Thin Sections
Use these thin-section slides with a polarizing microscope to view rock specimens in detail. Includes: biotite granite, olivine basalt, red sandstone, encrinal limestone, mica schist, augen gneiss, storage box and identification guide.
Note: collections are subject to change depending upon availability of individual items. Set of six.