Ward's® Economy Spectroscopes
Supplier: Avantor
PH0596CBTW
470104-988KT 170 CAD
470104-988 470105-358 470218-750
Ward's® Economy Spectroscopes
Spectrophotometers and Photometers UV/Visible Spectrophotometers
See the spectrum of various light sources with this economical device
  • Introduces students to many properties of visible light and atomic structure
  • Diffraction grating securely housed in end cap
  • Easily adjust orientation of the spectrum by twisting the slit end cap
  • Long tube provides dark background and easy viewing angle
The Wards® economy spectroscope has been designed to produce a vivid, bright spectrum of visible light decomposed by a diffraction grating. The resulting spectrum introduces students to many properties of visible light and atomicMore Product Information