Metal Cube and Slab Set
Supplier: American Scientific
CEM-225
470104-974EA 36.9 CAD
470104-974
Metal Cube and Slab Set
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
A steel cube, an aluminum cube and a rectangular aluminum slab have each been painted to conceal the identity of the metal. The steel and aluminum cubes have equal volume while the aluminum slab has greater volume. One plastic metric ruler is included with this set. For use with IPS textbook. Set of three.
WARNING: CHOKING HAZARD - Small parts. Not for children under 3 years.