Somso® Animal Cell Model
Supplier: MARCUS SOMMER
ZOS 120
470102-804EA 2256.1 CAD
470102-804
Somso® Animal Cell Model
Models Zoology Models
Highly detailed and brightly colored for easy identification.
  • Large size for easy organelle identification
  • 5 year manufacturers warranty
Enlarged 20,000 times, in SOMSO-Plast®. After Christian Groß, Head of Biology Department (retired). The model shows the recently discovered principles of the delicate structure of an animal cell.
In addition to the organelle like nucleus, endoplasmic reticulum, mitochondria, ribosomes respectively polysomes and Golgi apparatus, the model also shows centrioles, lysosomes and fat vacuoles.
The presentation of theMore Product Information