Somso® Herbaceous Monocot Stem Section Model
Supplier: MARCUS SOMMER
BOS 22/3
470029-848EA 1386.2 CAD
470029-848
Somso® Herbaceous Monocot Stem Section Model
Models Botany Models
View Details Of Corn Stem Up Close
  • Examine Stem Structure In Cross And Longitudinal Structure
  • Colors Similar To Typical Slide Staining For Easy Comparison
  • 5 Year Manufacturers Warranty
The colorful model of a corn stem section clearly illustrates monocot vascular structure. Enlarged 550X, the peripheral cross section of Zea mays details the xylem and phloem, tracheids, parenchyma, pith, and epidermis. Ideal for comparison to the structure of the Woody Dicot Stem Section Model.