GPI Anatomicals® Feline Heart and Lung Model
Supplier: GPI ANATOMICALS
9141
470029-470PK 151.2 CAD
470029-470 470029-471
GPI Anatomicals® Feline Heart and Lung Model
Models Zoology Models
Show The Effect Of Heartworm On Household Pets
Average feline heart and lungs infested with a single heartworm (Dirofilaria immitis). Cut-away view demonstrates structures in the heart (right ventricle, pulmonary trunk, pulmonary arteries) and lungs where adult-stage heartworms are normally found. The cut section of the lung indicates patches of heartworm-induced irritation.