Ward's® Cross-bedded Sandstone
Supplier: Avantor
470025-904EA 80.7 CAD
470025-904
Ward's® Cross-bedded Sandstone
Specimens Rock Specimens Specialty Rocks
Sawn and lacquered slab showing thinly bedded truncated layers in a well-cemented sandstone. Large hand specimen.
Order Now
SPECIFICATIONS
Environmentally Preferable