Secchi Transparency Tube
Supplier: Peco Sales
77292
470021-282EA 211.8 CAD
470021-282
Secchi Transparency Tube
Tubes Test Tubes
Estimate stream quality as it relates to suspended materials in the water.
  • Clear PVC
  • Tube is marked in centimeter graduations from 0 to 120 with a Secchi pattern at the bottom of the tube
To use, the tube is filled with water and drained using the drain tube until the Secchi pattern appears. The height of the water column can then be recorded.