Glass Tubing Cutter
Supplier: C & A Scientific
97-1601
470019-484EA 36.7 CAD
470019-484
Glass Tubing Cutter
Cutters
Made for easy glass cutting.
  • Cuts up to 11/2" dia. glass tubes
The spring loaded cutting wheel stays open until used.