Portable Fuming Chamber
Supplier: FORENSICS SOURCE
1-4676
470019-340EA 749.49 CAD
470019-340
Portable Fuming Chamber
Educational Materials Forensics Educational Materials Forensics Supplies
Bring Your Equipment to the Crime!
  • Made from durable ABS plastic
  • Includes (2) hanging bars with (4) clips for suspending evidence
  • Magnetic latch simplifies chamber loading and unloading
  • Convenient metal carrying handle allows for easy transportation and storage at the crime scene or the lab
The roomy chamber fits a wide variety of commonly processed evidence including cans, bottles and weapons. A special port incorporated in the design allows the attachment of an exhaust tube to vent fumes. This small but capable chamber can be used at home, in theMore Product Information