Full Spear Point Dissecting Needle
Supplier: Dunrite Instruments
141035
470018-970BX 439.25 CAD
470018-970
Full Spear Point Dissecting Needle
Dissection Needles
The spear point is useful for cutting fascia and cleaning tissue from small structures.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.