Science Take-Out® Kidney Donor
Supplier: SCIENCE TAKE-OUT, LLC
102 102U 102R 102-NOTG
470017-728EA 43.2 CAD
470017-728 470017-768 470017-808 470017-729
Science Take-Out® Kidney Donor
Educational Materials Biology Educational Materials Physiology Learning Activities
Which Person Would Make The Best Kidney Donor?
  • Available As A Single Kit With Materials For 1 Student Or Group
  • Unassembled 10 Pack For Additional Savings
  • No Additional Equipment Needed
Which family member can safely donate a kidney to a patient who needs a kidney transplant? Perform laboratory tests and analyze the results to determine who should donate a kidney to the patient. Conduct simulations of the following laboratory tests: urinalysis, blood typing, HLA tissue typing. Analyze donor survival data. This complete experiment setup contains allMore Product Information