CENCO® Coulomb and Current Apparatus
Supplier: Wards
2353A 2353
470016-192EA 231.25 CAD
470016-192 470016-194 470014-660
CENCO® Coulomb and Current Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
This apparatus is full of activities.
  • Sensitive Torque Arm
  • Straight Aluminum Rods
  • Instructions Included
  • Adjustable Counterweight
  • Optical Lever
The sensitive torque arm has as adjustable counterweight and an optical lever for measuring tiny forces with the null method. It comes with a pair of straight aluminum rods that measure the repulsive force between parallel, current-carrying conductors and a pair of plates for measuring the attractive force between charged capacitor plates. A current of five to 10A is required to cause sufficient deflection inMore Product Information