Four-Way Soil Analyzer
Supplier: Luster Leaf
1880
470013-572CS 34.15 CAD
470013-572
Four-Way Soil Analyzer
Analysers Elemental Analysers
With this analyzer, you can measure the most important plant-growing conditions.
  • No Batteries Required
  • Length: 4"/
  • Width: 2½"1/4
  • Diameter: 1¼"/
  • Functions: Analyzing Fertilizer, pH, Moisture, and More
To take measurements with this analog meter, insert it in the soil and move the switch to the factor you want to test. It will measure the amount of fertilizer (too little, ideal, too much), light (0–10), moisture (1–4), and pH (0–9.0).
This meter does not require batteries.