Ward's® Voltage and Currents Lab Activity
Supplier: Science First
14476
470013-324KT 409.6 CAD
470013-324
Ward's® Voltage and Currents Lab Activity
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Explore Direct Current Circuits
Examine the effects varying voltage has on common components in simple, DC circuits. See how a standard 6 V light bulb varies in brightness, temperature, and color at different voltages, and the changes in intensity and blinking rate in a 2.5 V “blinky” bulb. You can also see how voltage and circuit polarity effect the operation and intensity of an LED. An optional investigation introduces circuit diagrams and the relationship between current and voltage. It includes enough bulbs, connectors, LEDs, and more for five setups, as well as a teacher’s guide andMore Product Information