American Scientific Unmounted Spherical Bi-Concave Lens
Supplier: American Scientific
3309-07
470013-320EA 5.8 CAD
470013-320
American Scientific Unmounted Spherical Bi-Concave Lens
Educational Materials Physics Educational Materials Optics Learning Activities
Used for reducing the image size or spreading light out.
  • Bi-concave shape: Spherical design with both surfaces curving inward for diverging light
  • Unmounted: No mounting frame, allowing for flexible integration into various setups
  • 38 mm diameter: Specific diameter suitable for diverse optical applications
  • 300 mm focal length: Long focal length ideal for precise light manipulation and beam expansion
  • Cost-effective: Unmounted lenses can be more affordable and versatile compared to mounted alternatives
The unmounted spherical bi-concave lens is aMore Product Information