Ward's® Chemistry Density Column Demonstration
Supplier: Avantor
470013-198EA 127 CAD
470013-198
Ward's® Chemistry Density Column Demonstration
Educational Classroom Kits and Activities
Interactive Illustration of Density Principles
  • The perfect supplement to your chemistry lectures
  • Can be performed in just 10–30 minutes.
  • Requires only basic labware that you provide
In this “learn-by-doing” activity, you will prepare a column of five liquids, each of which has a different density, and will then compare the density of various small, solid objects by dropping them into the “density column.”
Order Now
SPECIFICATIONS
Environmentally Preferable