Genecon Hand Operated Generator
Supplier: NADA SCIENTIFIC LTD.
N99-B-2637-002 N99-B10-2632
470012-820EA 59.55 CAD
470012-820 470006-386
Genecon Hand Operated Generator
Batteries
With this generator, students have the power of electricity in the palm of their hands.
  • Tactile Fun for Students
  • Transparent Design for Internal Viewing
  • Students Learn about Generators and Motors
  • Color-Coded Cord and Plug
  • Generator Produces 12V
The modern design offers an unprecedented 12VDC of generating capacity. The transparent ABS resin construction lets users see the internal structure of the generator. The new, color-coded cord and plug have been thoughtfully designed for durability and long-lasting performance.
The direction of the rotationMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable