How Does All That DNA Fit? Lab Activity
Supplier: Avantor
470012-256EA 126.55 CAD
470012-256
How Does All That DNA Fit? Lab Activity
Educational Classroom Kits and Activities
Apply skills of dimensional analysis and exponential notation.
Discover startling conclusions about the linear quantity of DNA contained within a single cell and within the entire human body. In this engaging activity, students use algebra to calculate the length of DNA, as well as to determine a compaction ratio. They then pack thread into capsules to simulate chromatin packing in a nucleus. These determinations illustrate the effect and importance of molecular packing or supercoiling in living cells. A final challenge applies the skills of dimensional analysis and exponential notationMore Product Information