Introduction to Physical Properties of Proteins Lab Activity
Supplier: Kemtec
8-152
470008-064KT 425 CAD
470008-064
Introduction to Physical Properties of Proteins Lab Activity
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Provide a Thorough Overview of the Function of Proteins
Students will discover how physical parameters, such as temperature, pH, salt concentration and dielectric constants affect the isolation and purification of proteins. They will conduct experiments in which they will precipitate casein out of milk, separate albumin and gelatin through fractionation, determine the effects of acetone on albumin and gelatin, denature albumin, and precipitate protein with lead acetate. It includes enough materials for eight setups, a teacher’s guide, and student data sheets.